missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EasySep™ Human Whole Blood CD34 Positive Selection Kit II

Product Code. 16605645
Encompass
Change view
Click to view available options
Quantity:
75 mL Whole Blood (37 mL Buffy Coat)
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16605645 75 mL Whole Blood (37 mL Buffy Coat) 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16605645 Supplier STEMCELL Technologies Supplier No. 17879

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Immunomagnetic positive selection of human CD34+ cells from whole blood or buffy coat

  • The EasySep™ Human Whole Blood CD34 Positive Selection Kit II is designed to isolate CD34+ cells from fresh whole blood or freshly prepared buffy coat by positive selection
  • Desired cells are targeted with Tetrameric Antibody Complexes recognizing CD34 and dextran-coated magnetic particles
  • The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding
  • Labeled cells are separated using an EasySep™ magnet without the use of columns
  • Cells of interest remain in the tube while unwanted cells are poured off
  • This product can be used instead of the EasySep™ Human Whole Blood CD34 Positive Selection Kit (Catalog #18086)
  • If isolating CD34+ cells from fresh cord blood, we recommend using the EasySep™ Human Cord Blood CD34 Positive Selection Kit II (Catalog #17896)
  • Alternatively, if isolating CD34+ cells from any other sample type, the EasySep™ Human CD34 Positive Selection Kit II (Catalog #17856) should be used

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Isolation
Quantity 75 mL Whole Blood (37 mL Buffy Coat)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.