Learn More
STEMCELL Technologies EasySep™ Direct Human Total Lymphocyte Isolation Kit
Product Code. 17147941
Encompass
Change view
Click to view available options
Quantity:
100 mL whole blood
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17147941 100 mL whole blood 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17147941 Supplier STEMCELL Technologies Supplier No. 19655
Immunomagnetic isolation of total lymphocytes directly from whole blood
  • The EasySep™ Direct Human Total Lymphocyte Isolation Kit isolates highly purified lymphocytes directly from human whole blood by immunomagnetic negative selection
  • Isolated cells are untouched and available for direct use in flow cytometry, culture​,​ DNA/RNA extraction or other downstream applications​
Spezifikation
Cell Type Lymphocytes
For Use With (Application) Cell Isolation
Quantity 100 mL whole blood