Learn More
STEMCELL Technologies EasySep™ Direct Human Neutrophil Isolation Kit
Product Code. 15805912
Encompass
Change view
Click to view available options
Quantity:
100 mL whole blood
Unit Size:
Each
Product Code. Quantity unitSize
15805912 100 mL whole blood Each
1 options
Product Code. 15805912
Brand: STEMCELL Technologies 19666
Immunomagnetic isolation of neutrophils directly from whole blood
  • The EasySep™ Direct Human Neutrophil Isolation Kit isolates functional, highly purified neutrophils directly from human whole blood by immunomagnetic negative selection
  • No lysis, density gradient centrifugation or other processing steps are required and isolat​ed cells are immediately available for flow cytometry, functional assays, culture, and other downstream applications
  • This product replaces the EasySep™ Human Neutrophil Isolation Kit (Catalog #19257) for even faster cell isolations
Specifications
Cell Type Granulocytes and Subsets
For Use With (Application) Cell Isolation
Quantity 100 mL whole blood