missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DUSP22 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16514-1-AP-20UL
This item is not returnable.
View return policy
Description
Mitogen-activated protein (MAP) kinases are a large class of proteins involved in signal transduction pathways that are activated by a range of stimuli and mediate a number of physiological and pathological changes in the cell. Dual specificity phosphatases (DSPs) are a subclass of the protein tyrosine phosphatase (PTP) gene superfamily, which are selective for dephosphorylating critical phosphothreonine and phosphotyrosine residues within MAP kinases. DSP gene expression is induced by a host of growth factors and/or cellular stresses, thereby negatively regulating MAP kinase superfamily members including MAPK/ERK, SAPK/JNK and p38. DUSP22 dephosphorylates ERK2 MAP kinase and JNK. DUSP22 displays highest in thymus, but it is also detectable in monocytes and lymphocytes.Specifications
| DUSP22 | |
| Polyclonal | |
| Unconjugated | |
| DUSP22 | |
| DUSP22, JKAP, JNK stimulatory phosphatase 1, JSP 1, JSP1, LMW DSP2, LMWDSP2, MAP kinase phosphatase x, MKP x, MKPX, VHX | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 105352, 56940 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q99N11, Q9NRW4 | |
| DUSP22 | |
| DUSP22 Fusion Protein Ag9734 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction