Learn More
Applied Biosystems™ DS-33 GeneScan™ Installation Standards with GeneScan™ 600 LIZ™ Size Standard v2.0
Product Code. 16380824 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
Product Code. Quantity unitSize
16380824 1 kit 1 set
1 options
Product Code. 16380824
Brand: Applied Biosystems™ 4376911
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
Each kit contains four tubes of GeneScan™ Installation Standard DS-33 and one tube of GeneScan™ 600 LIZ™ size standard v2.0.
The DS-33 GeneScan™ Installation standard consists of pooled PCR products labeled with 6-FAM™, VIC™, NED™ and PET™ dyes.
The kit also contains a tube of GeneScan™ 600 LIZ™ Size Standard 2.0.
Order Info
Kit
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Each kit contains four tubes of GeneScan™ Installation Standard DS-33 and one tube of GeneScan™ 600 LIZ™ size standard v2.0.
Store the DS-33 tubes at -25° to -15°C
Store the GeneScan 600 LIZ Size Standard v2.0 opon thawing at 2@deg; to 8°C, protect from light.
Format Tube
For Use With (Equipment) 3730xl DNA Analyzer, SeqStudio Genetic Analyzer, 3500 Genetic Analyzer, 3730 DNA Analyzer, 3500xL Genetic Analyzer
Label or Dye FAM, LIZ, NED, PET, VIC
Product Line GeneScan, LIZ
Product Type Installation Standard
Quantity 1 kit
Shipping Condition Dry Ice
Test Time Standard