Learn More
Fisherbrand™ PTFE Magnetic Stir Bar, Double-sided crosshead
Product Code. 10211812 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric):
10 mm
14 mm
17 mm
22 mm
30 mm
35 mm
40 mm
60 mm
Unit Size:
Each
Pack of 10
Pack of 5
Product Code. 10211812
Only null left
Add to basket
Add to basket
Designed for use with tube-like containers yet very effective as general stirrers. The PTFE-coated magnet stirrers include a crosshead on both sides of the stirrer for greater turbulence.
  • Stable, general-purpose magnetic stirrer
  • Designed for use with tube-like containers yet very effective as general stirrers
  • Double-sided crossheads provider greater turbulence during mixing
  • Autoclavable
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Height (Metric) 15mm
Coating PTFE
Diameter (Metric) 60 mm
Quantity 2/Pk.
Type PTFE Stirrer Bars Crosshead