Learn More
Miele Professional™ Dosing System DOS K 85
Shop All Miele Products
Click to view available options
Includes:
5 to 10 L Canister
Unit Size:
1 sheet
Description
- Dispenser monitoring high process security as per EN ISO 15883
- For dispensing liquid cleaning agent/neutralising agent
- 352 mm suction lance for 5 - 10 L canisters
- Siphon conversion kit for 10 - 30 L media canisters available
Specifications
Specifications
| Product Type | Dosing Pump |
| Includes | 5 to 10 L Canister |
| Depth (English) | 18 7/8 in. |
| Depth (Metric) | 47.80 cm |
| Dimensions (L x W x H) | 47.80 x 19.80 x 16.00 cm |
| Height (English) | 6 5/16 in. |
| Height (Metric) | 16.00 cm |
| Length (English) | 18 7/8 in. |
| Length (Metric) | 47.80 cm |
| Width (English) | 7 13/16 in. |
| Show More |