Learn More
Miele Professional™ Dosing system
Shop All Miele Products
Click to view available options
For Use With (Equipment):
Washer-disinfectors, laboratory
Unit Size:
Each
Description
- Compatible with all PG 8535
- Adjustable via electronic control of the machine
- Length of the suction pipe 300 mm for 5 and 10 l cans
- Length of the guide cable: 1.90 m
Specifications
Specifications
| Product Type | Dispensing System |
| Model | DOS G-60 Dispenser Module |
| For Use With (Equipment) | Washer-disinfectors, laboratory |
| Quantity | 1 Each |