Learn More
Memmert™ Door-Open-Recognition for UN110plus
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UN110plus Oven
Unit Size:
Each
Description
- Only for units with TwinDISPLAY
Specifications
Specifications
| For Use With (Equipment) | UN110plus Oven |
| Type | Door Open Recognition |