Learn More
MP Biomedicals Dog Diet, Purified, Powder
Product Code. 11402811 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 kg
20 kg
50 kg
Unit Size:
10kg
20kg
50kg
Product Code. 11402811
Brand: MP Biomedicals undefined
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Dog diet containing casein, sucrose, lard, fiber, mineral and vitamin mix.
  • Diet specifically made for dogs and dog research
  • Dog diet containing casein, sucrose, lard, fiber, mineral and vitamin mix
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Autoclavable
Product Type Animal Diet
Disposable Single-use
Format Powder
Quantity 10 kg
Sterility Non-sterile
Storage Requirements Room Temperature
Length (English) 1 ft.
Length (Metric) 0.3048 m
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.