missing translation for 'onlineSavingsMsg'
Learn More

DLK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15680-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16884954

  • 3468.15 NOK / 150µL
Estimated Shipment: 20-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Regulates adipogenesis.
Specifications

Specifications

DLK2
Polyclonal
Unconjugated
DLK2
DLK 2, DLK2, EGF like protein 9, EGFL9, Protein delta homolog 2
Rabbit
Antigen Affinity Chromatography
RUO
106565, 316232, 65989
-20°C
Liquid
Western Blot, Immunoprecipitation
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
D3ZUK3, Q6UY11, Q8K1E3
Dlk2
DLK2 Fusion Protein Ag8170
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers