missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLK2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | DLK2 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16894954
|
Proteintech
15680-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16884954
|
Proteintech
15680-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 17-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Regulates adipogenesis.Specifications
DLK2 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
D3ZUK3, Q6UY11, Q8K1E3 | |
106565, 316232, 65989 | |
DLK2 Fusion Protein Ag8170 | |
Primary | |
-20°C | |
DLK2 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
DLK 2, DLK2, EGF like protein 9, EGFL9, Protein delta homolog 2 | |
Dlk2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |