missing translation for 'onlineSavingsMsg'
Learn More

DL-Methionine ethyl ester hydrochloride, 98+%, Thermo Scientific™

Product Code. 11421140
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11421140 100 g 100g
11401140 5 g 5g
11411140 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11421140 Supplier Thermo Scientific Alfa Aesar Supplier No. H25781.22
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6297-53-6
Molecular Formula C7H16ClNO2S
Molecular Weight (g/mol) 213.72
MDL Number MFCD00237859
InChI Key KPWCQEUBJAIORR-UHFFFAOYNA-N
Synonym dl-methionine ethyl ester hydrochloride, h-met-oet hcl, c7h15no2s.hcl, h-dl-met-oet hcl, h-dl-met-oet.hcl, dl-methionine ethyl ester hcl, ethyl 2-amino-4-methylthiobutanoate, chloride, ethyl 2-azanyl-4-methylsulfanyl-butanoate hydrochloride, 2-amino-4-methylthio butanoic acid ethyl ester hydrochloride, ethyl 2-amino-4-methylsulfanyl butanoate hydrochloride
PubChem CID 12641634
SMILES Cl.CCOC(=O)C(N)CCSC

Specifications

Melting Point 72°C to 75°C
IUPAC Name ethyl 2-amino-4-(methylsulfanyl)butanoate hydrochloridyl
Quantity 100 g
Formula Weight 213.73
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material DL-Methionine ethyl ester hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.