accessibility menu, dialog, popup

UserName

CAS RN 6297-53-6

IUPAC Name: ethyl 2-amino-4-(methylsulfanyl)butanoate hydrochloridyl
Synonyms: ethyl 2-amino-4-(methylsulfanyl)butanoate hydrochloridyl
Molecular Weight (g/mol): 212.71
Molecular Formula: C7H15ClNO2S
InChi Key: YDVXNFFYLFHDLB-UHFFFAOYNA-N
SMILES: [Cl].CCOC(=O)C(N)CCSC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results