Learn More
Hanna Instruments™ Dissolved Oxygen Electrolyte Solutions
298.00 NOK - 351.00 NOK
Specifications
Capacity (Metric) 30 mL
Type Dissolved Oxygen Electrolyte Solution
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
10643272
View Documents Promotion Details
Hanna Instruments™
HI7041S
Hanna's Polarographic Dissolved Oxygen Probes
298.00 NOK
30mL
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Add to basket
11534347
View Documents Promotion Details
Hanna Instruments™
HI7042S
Hanna's Galvanic Dissolved Oxygen Probes
351.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 21-05-2024
Specifications
30 mL
Dissolved Oxygen Electrolyte Solution