Learn More
Fisherbrand™ Benchtop Single Dispensing Bin
Product Code. 11314065 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color:
Neon Blue
Dimensions (L x W x H):
155 x 155 x 170 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Dimensions (L x W x H) Unit Size
11314065 Neon Blue 155 x 155 x 170 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11314065
Stock Up and Save Up to 60%
Use promo code 28009
View all promotions
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket
Access your lab materials with ease, even with gloved hands. Benchtop Single Dispensing Bin is ergonomically designed with a large, angled opening and an oversized lip.
Easily access your lab materials, even with gloved hands. Ideal for holding many different laboratory items including micro-centrifuge tubes.
Ergonomically designed:
  • Large, angled openings.
  • Oversized lip.
  • Lid remains open when tilted back.
Ideal for benchtop and wall mounted use:
  • Nonskid rubber feet.
  • Pre-drilled holes and included screws for wall mounting.
Specifications
Type Dispensing Bin
Material PMMA
Color Neon Blue
Shape Semi-circular
Quantity 1 Ea.
Dimensions (L x W x H) 155 x 155 x 170 mm
Length (English) 6.1 in.
Length (Metric) 155 mm
Width (Metric) 155 mm
Height (Metric) 170 mm