Learn More
Fisherbrand™ Catalyst PharMed BPT Formulation Pump Tubing
Product Code. 15531305 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
0.8 mm
1.6 mm
3.2 mm
4.8 mm
6.4 mm
8 mm
Unit Size:
Each
Product Code. 15531305
Only null left
Add to basket
Add to basket
Available in standard 15m Lengths.
    • Includes platinum-cured silicone, two TPEs and long-life PVC tubing material
    • Available in both thin and thick wall configurations
Specifications
Description Catalyst pump tubing
Diameter (Metric) Inner 8 mm
Thickness (Metric) 1.6 mm