Learn More
Thermo Scientific™ Dionex™ Fused Silica Tubing
Product Code. 11382123 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inner:
10 μm
20 μm
25 μm
30 μm
50 μm
75 μm
100 μm
150 μm
200 μm
Length (Metric):
2.5 m
5 m
Unit Size:
Each
Product Code. Diameter (Metric) Inner Length (Metric) unitSize
11382123 200 μm 5 m Each
11322123 20 μm 5 m Each
11332123 25 μm 5 m Each
11342123 50 μm 5 m Each
11352123 75 μm 5 m Each
11362123 100 μm 5 m Each
11372123 150 μm 5 m Each
11312143 10 μm 5 m Each
11312233 30 μm 2.5 m Each
9 options
Product Code. 11382123
Brand: Thermo Scientific™ 160481
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Thermo Scientific™ Fused Silica Tubing provides excellent chemical durability and inertness.
Thermo Scientific™ Fused Silica Tubing provides excellent chemical durability and inertness.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) LC-Packings Ultimate Capillary/Nano HPLC
Material Fused Silica
Type Capillary Tubing
Length (Metric) 5 m
Diameter (Metric) Inner 200 μm
Diameter (Metric) Outer 280 μm
Product Line nanoLC