Learn More
Diisobutyl adipate, 97%, Thermo Scientific™
Product Code. 11429466
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
Product Code. 11429466
Brand: Thermo Scientific Alfa Aesar A19186.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 141-04-8
Molecular Formula C14H26O4
Molecular Weight (g/mol) 258.358
MDL Number MFCD00053722
InChI Key RDOFJDLLWVCMRU-UHFFFAOYSA-N
Synonym diisobutyl adipate, isobutyl adipate, diba, ftaflex diba, plasthall diba, bis 2-methylpropyl hexanedioate, hexanedioic acid, bis 2-methylpropyl ester, adipic acid, diisobutyl ester, diisobutyl hexanedioate, adipic acid bis 2-methylpropyl ester
PubChem CID 8831
ChEBI CHEBI:34708
IUPAC Name bis(2-methylpropyl) hexanedioate
SMILES CC(C)COC(=O)CCCCC(=O)OCC(C)C
Specifications
Density 0.954
Boiling Point 293°C
Flash Point >110°C (230°F)
Refractive Index 1.432
Quantity 25 g
Formula Weight 258.36
Percent Purity 97%
Chemical Name or Material Diisobutyl adipate