accessibility menu, dialog, popup

UserName

CAS RN 141-04-8

H3CCH3OOOOCH3CH3

CAS RN 141-04-8

IUPAC Name: 1,6-bis(2-methylpropyl) hexanedioate
Synonyms: DIBA ftaflex diba plasthall diba
Molecular Weight (g/mol): 258.36
Molecular Formula: C14H26O4
InChi Key: RDOFJDLLWVCMRU-UHFFFAOYSA-N
SMILES: CC(C)COC(=O)CCCCC(=O)OCC(C)C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)