Learn More
Digi-Sense™ Advanced Precalibrated RTD Thermometers
Product Code. 15890056
Change view
Click to view available options
Model:
Data logging
Not data logging
Probe Type:
2-input
Single input
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Probe Type unitSize
15890056 Data logging 2-input Each
15622617 Not data logging Single input Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15890056 Supplier Digi-Sense™ Supplier No. 2025096
Add to basket
Add to basket
Includes
Protective rubber armor boot with built-in stand, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal. Data logging models 20250-92, -94, and -96 also include a USB cable and software.
Perfect for the lab to production floor
  • Reliable measurements, guaranteed!
  • NIST-traceable certificate included
  • Heavy duty construction to protect against knocks and jars.
  • Super lager LCD for easy viewing of data
  • Accepts a wide variety of probes
  • Min, Max, and Avg readings, for quick analysis of the object
  • Electronic Offset function allows compensation of probe errors
Warranty
2 Years
Specifications
Disposable Reusable
Height (English) 1 in.
Height (Metric) 2.6 cm
Includes Protective rubber armor boot with built-in stand, one 9 V battery, and NIST-traceable calibration report with data supplied by InnoCal. Data logging model 20250-96 also includes\ a USB cable and software.
Length (Metric) 178 mm
Material Plastic
Measurement Time <1 sec.
Model Data logging
Probe Type 2-input
Resolution 0.1°F (0.1°C) from 148 to 1562°F (-99.9 to 850°C); 1°F (1°C) outside range
Width (English) 2.25 in.
Product Type RTD Thermometer
Data Storage 16,000 Records
Length (English) 7 in.
Max. Temperature (English) 1562°F
Max. Temperature (Metric) 850°C
Min. Temperature (English) -320°F
Min. Temperature (Metric) -200°C
Temperature Range (English) -320°F to +1562°F
Temperature Range (Metric) -200°C to +850°C
Calibrated NIST-traceable
For Use With (Equipment) Pt 100 ohm RTD Probes
Battery Type 9V
Accuracy ±1.5°F (0.8°C) from -148 to 392°F (-99.9 to 199.9°C); ±2°F (1°C) ±0.5% of reading outside range
Display Backlit LCD
Width (Metric) 5.7 cm