Learn More
Diclofenac diethylamine, MedChemExpress
Description
Diclofenac diethylamine is a potent and nonselective anti-inflammatory agent, acts as a COX inhibitor, with IC50s of 4 and 1.3 nM for human COX-1 and COX-2 in CHO cells, and 5.1 and 0.84 μM for ovine COX-1 and COX-2, respectively. Diclofenac diethylamine induces apoptosis of neural stem cells (NSCs) via the activation of the caspase cascade.
Specifications
Specifications
| Chemical Name or Material | Diclofenac diethylamine |
| CAS | 78213-16-8 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H22Cl2N2O2 |
| Quantity | 5 g |
| Solubility Information | DMSO : 150 mg/mL (406.18 mM; Need ultrasonic) |
| SMILES | O=C(O)CC1=CC=CC=C1NC2=C(Cl)C=CC=C2Cl.CCNCC |
| Molecular Weight (g/mol) | 369.29 |
| Formula Weight | 369.29 |
| Show More |