Learn More
VEE GEE Dial Thermometer
Dial thermometers of varying sizes based on personal preferences.
85.05 NOK - 1435.00 NOK
Specifications
Material | Metal |
---|---|
Product Type | Serialized Glass Thermometer |
Accuracy | ±1 subdivision |
Product Code | Brand | Display Dimensions | Immersion Length | Length (English) Probe | Temperature Range (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Display Dimensions | Immersion Length | Length (English) Probe | Temperature Range (English) | Price | Quantity & Availability | ||
17861224
|
VEE GEE
81070 |
2.54 cm Diameter | 12.7 cm | 5 in. | -4 to +158°F |
85.05 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17801204
|
VEE GEE
81110 |
2.54 cm Diameter | 12.7 cm | 5 in. | 14 to 316.4°F |
85.05 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17811254
|
VEE GEE
81125 |
2.54 cm Diameter | 12.7 cm | 5 in. | 25 to 125°F |
85.05 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17821234
|
VEE GEE
81550 |
2.54 cm Diameter | 12.7 cm | 5 in. | 50 to 550°F |
85.05 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17831254
|
VEE GEE
82100-36 |
6.62 cm Diameter | 91.4 cm | 36 in. | 32 to 212°F |
1190.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17841234
|
VEE GEE
82110 |
4.5 cm Diameter | 20.3 cm | 36 in. | 14 to 230°F |
130.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17861234
|
VEE GEE
82125 |
4.5 cm Diameter | 20.3 cm | 48 in. | 25 to 125°F |
130.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17861214
|
VEE GEE
82160-12 |
7.6 cm Diameter | 30.5 cm | 6 in. | -40 to +160°F |
391.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17821204
|
VEE GEE
82160-6 |
7.6 cm Diameter | 15.2 cm | 12 in. | -40 to +160°F |
294.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17841254
|
VEE GEE
82200-36 |
6.62 cm Diameter | 91.4 cm | 8 in. | 0 to 200°F |
1190.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17851254
|
VEE GEE
82200-48 |
6.62 cm Diameter | 122 cm | 8 in. | 0 to 200°F |
1435.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17801244
|
VEE GEE
82220 |
4.5 cm Diameter | 20.3 cm | 8 in. | 0 to 200°F |
130.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17891234
|
VEE GEE
82550 |
4.5 cm Diameter | 20.3 cm | 8 in. | 50 to 550°F |
130.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
17811194
|
VEE GEE
82550DG |
5.1 cm Diameter | 20.3 cm | 8 in. | 50 to 550°F |
169.00 NOK
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
||
Description
- The dial thermometer shows the temperature using a metal stem and a circular scale.
- It doesn't need batteries, it's tough, and you can change the readings by using the calibration screw on the back.
Specifications
Metal | |
Serialized Glass Thermometer | |
±1 subdivision |