missing translation for 'onlineSavingsMsg'
Learn More

DHODH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14877-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16893754

  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene catalyzes the fourth enzymatic step, the ubiquinone-mediated oxidation of dihydroorotate to orotate, in de novo pyrimidine biosynthesis. This protein is a mitochondrial protein located on the outer surface of the inner mitochondrial membrane.
Specifications

Specifications

DHODH
Polyclonal
Unconjugated
DHODH
DHOdehase, DHODH, dihydroorotate dehydrogenase, Dihydroorotate oxidase
Rabbit
Antigen Affinity Chromatography
RUO
1723, 56749, 65156
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O35435, Q02127, Q63707
DHODH
DHODH Fusion Protein Ag6649
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers