missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DHODH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | DHODH |
|---|---|
| Concentration | 0.25 mg/mL |
| Applications | Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16803764
|
Proteintech
14877-1-AP-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16893754
|
Proteintech
14877-1-AP-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene catalyzes the fourth enzymatic step, the ubiquinone-mediated oxidation of dihydroorotate to orotate, in de novo pyrimidine biosynthesis. This protein is a mitochondrial protein located on the outer surface of the inner mitochondrial membrane.Specifications
| DHODH | |
| Immunoprecipitation, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Rat, Human, Mouse | |
| O35435, Q02127, Q63707 | |
| 1723, 56749, 65156 | |
| DHODH Fusion Protein Ag6649 | |
| Primary | |
| -20°C | |
| DHODH |
| 0.25 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| DHOdehase, DHODH, dihydroorotate dehydrogenase, Dihydroorotate oxidase | |
| DHODH | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title