Specifications
| Light Source | UV |
| Wavelength | 400/185 nm |
| Aperture Size (Metric) | 1 mm |
| For Use With (Equipment) | Shimadzu UV Spec,BioSpec-mini |
| Packaging Quantity | 1 Ea. |
| Product Type | Deuterium Lamp |