missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoplakin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25318-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody detects both DP1 (280-330 kDa) and DP2 (240-260 kDa).
Desmoplakin (DSP) is a major high molecular weight protein of the desmosomes. It is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes. Desmoplakin mutations are associated with Keratoderma and cardiomyopathy.Specifications
Desmoplakin | |
Polyclonal | |
Unconjugated | |
DSP | |
desmoplakin, Desmoplakin I, Desmoplakin II, DP, DPI, DPII, DSP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
109620, 1832 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.4 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
E9Q557, P15924 | |
DSP | |
Desmoplakin Fusion Protein Ag18065 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |