missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Desmoplakin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | Desmoplakin |
---|---|
Concentration | 0.4 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803915
|
Proteintech
25318-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16893905
|
Proteintech
25318-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This antibody detects both DP1 (280-330 kDa) and DP2 (240-260 kDa).
Desmoplakin (DSP) is a major high molecular weight protein of the desmosomes. It is involved in the organization of the desmosomal cadherin-plakoglobin complexes into discrete plasma membrane domains and in the anchoring of intermediate filaments to the desmosomes. Desmoplakin mutations are associated with Keratoderma and cardiomyopathy.Specifications
Desmoplakin | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human | |
E9Q557, P15924 | |
109620, 1832 | |
Desmoplakin Fusion Protein Ag18065 | |
Primary | |
-20°C | |
DSP |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
desmoplakin, Desmoplakin I, Desmoplakin II, DP, DPI, DPII, DSP | |
DSP | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |