missing translation for 'onlineSavingsMsg'
Learn More

Deoxyribonucleic acid, MP Biomedicals™

Product Code. 11428531 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428531 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11428531 Supplier MP Biomedicals Supplier No. 0215227580

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 99675-55-5
Synonym DNA sodium salt, Desoxiribon

Specifications

Color White
Quantity 100 g
Formula Weight ≈6,000,000
Physical Form Powder
Chemical Name or Material Deoxyribonucleic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.