CAS RN 99675-55-5
CAS RN 99675-55-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Deoxyribonucleic acid, MP Biomedicals™
Deoxyribonucleic acid, MP Biomedicals™
CAS: 99675-55-5 Synonym: DNA sodium salt,Desoxiribon