Learn More
Eppendorf™ Delivery to Factory Gate
Product Code. p-7221201
Click to view available options
For Use With (Application):
Delivery of >100, <180 kg Equipment
Delivery of >180 kg Equipment
Delivery of <100 kg Equipment
Unit Size:
Each
Product Code. 16450124
Brand: Eppendorf™ 2277000009
Delivery to the factory gate or first door at the end customer side
  • Delivery to the factory gate
  • Delivery to first door at the end customer side
TRUSTED_SUSTAINABILITY
Specifications
Description Delivery to factory gate (<100 kg)
Service Offering Factory Gate Delivery
For Use With (Application) Delivery of <100 kg Equipment