Learn More
Eppendorf™ Delivery to Factory Gate
Product Code. 17141066
Change view
Click to view available options
For Use With (Application):
Delivery of >100, <180 kg Equipment
Delivery of >180 kg Equipment
Delivery of <100 kg Equipment
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Application) Unit Size
17141066 Delivery of >100, <180 kg Equipment Each
16450124 Delivery of <100 kg Equipment Each
17151066 Delivery of >180 kg Equipment Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17141066 Supplier Eppendorf™ Supplier No. 2277000010
 Restricted Item
Estimated Shipment: 07-12-2026
Delivery to the factory gate or first door at the end customer side
  • Delivery to the factory gate
  • Delivery to first door at the end customer side
TRUSTED_SUSTAINABILITY
Specifications
Product Type Factory Gate Delivery
For Use With (Application) Delivery of >100, <180 kg Equipment