Learn More
Fisherbrand™ Deionization Cartridges
Product Code. 11954159 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Color Indicator:
Blue cartridge turns ocher brown
Green cartridge turns purple
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Indicator Unit Size
11954159 Blue cartridge turns ocher brown Each
11944159 Green cartridge turns purple Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11954159
  • ONLINE EXCLUSIVE PRICE
    885.00 NOK
    833.81 NOK / Pack of 1
    Save 51.19 NOK
    5% Off
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
Purify water with single use cartridges. Fisherbrand™ Deionization Cartridges change color when saturation is detected.
  • Detects the saturation by color change
  • Direct connection to the water network
  • A wide choice of cartridges to meet all needs with reduced cost
  • No electrical connection
  • A transparent building to monitor the saturation of the resin without auxiliary measurement
  • The disposable cartridges contain only new resins. The performance level is constant and guaranteed
  • At saturation of the resin cartridges are simply discarded
  • The small dimensions allow easy installation
  • Applications: dishwashing, cooling circuit filling, battery, battery and radiator antifreeze preparation, photo development
Specifications
Color Indicator Blue cartridge turns ocher brown
Capacity Depends on the quality of the inlet water
Flow Rate 20 L/hr.
Resin Type Ion Exchanger
Filter Size 500 mm
For Use With (Application) For water purification, sensitive applications
For Use With (Equipment) For R3 series
Pressure 3 bar
Quantity 1 Each
Temperature (Metric) Max. Operating 35°C
Temperature (Metric) Operating 10°C to 35°C
Type Cartridge