Learn More
Fisherbrand™ Decapping Heads for Pneumatic Hand-held Crimping Tool
Product Code. p-7113876 Shop All Fisherbrand Products
Change view
Click to view available options
Size:
11 mm
13 mm
20 mm
32 mm
Unit Size:
Each
Product Code. Size unitSize
11889440 32 mm Each
11854970 13 mm Each
11854890 11 mm Each
11885050 20 mm Each
4 options
Product Code. 11889440
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with Pneumatic Hand-held Crimping Tool
  • Decapping Head
  • Pneumatic
  • Steel
TRUSTED_SUSTAINABILITY
Specifications
Description 32mm Decapping Head
Type Decapping Head
For Use With (Equipment) Aluminum Cap
Size 32 mm