Learn More
Fisherbrand™ Decapping Heads for Pneumatic Hand-held Crimping Tool
For use with Pneumatic Hand-held Crimping Tool
Brand: Fisherbrand™
6726.00 NOK valid until 2024-12-31
Use promo code "23262" to get your promotional price.
Additional Details : Weight : 0.34000kg
Description
- Decapping Head
- Pneumatic
- Steel
Specifications
Aluminum Cap | |
20mm Decapping Head |