missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress DBCO-(PEG)3-VC-PAB-MMAE
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Specifications
Specifications
| Molecular Weight (g/mol) | 1613.97 g/mol |
| Content And Storage | -20°C, stored under nitrogen ∣ The compound is unstable in solutions, freshly prepared is recommended. |
| Shipping Condition | Room temperature |
| Quantity | 10 mg |
| Solubility | DMSO : 60 mg/mL (37.18 mM; Need ultrasonic) |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction