missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Fructose-1,6-diphosphate dicalcium salt, 95%

Product Code. 15487849
Click to view available options
Quantity:
25 g
100 g
250 g
Unit Size:
100g
250g
25g
This item is not returnable. View return policy

Product Code. 15487849

Brand: Thermo Scientific Chemicals J66477.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6055-82-9 | C6H10Ca2O12P2 | 416.238 g/mol

D-Fructose-1,6-diphosphate dicalcium salt is essential for glycolysis to occur efficiently. It links to adenine nucleotides which regulate 6-phosphofructokinases (Pfks) that catalyze one of the rate limiting steps of glycolysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Fructose-1,6-diphosphate dicalcium salt is essential for glycolysis to occur efficiently. It links to adenine nucleotides which regulate 6-phosphofructokinases (Pfks) that catalyze one of the rate limiting steps of glycolysis.

Solubility
Soluble in water

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6055-82-9
Molecular Formula C6H10Ca2O12P2
Molecular Weight (g/mol) 416.238
MDL Number MFCD00135875
InChI Key MKSZTTCFYSGQHA-UHFFFAOYSA-J
Synonym d-fructose, 1,6-bis dihydrogen phosphate , calcium salt 1:2, dicalcium 2,3,4-trihydroxy-5-oxo-6-phosphonatooxyhexyl phosphate, dicalcium 2,3,4-tris oxidanyl-5-oxidanylidene-6-phosphonatooxy-hexyl phosphate
PubChem CID 53398690
IUPAC Name dicalcium;(2,3,4-trihydroxy-5-oxo-6-phosphonatooxyhexyl) phosphate
SMILES C(C(C(C(C(=O)COP(=O)([O-])[O-])O)O)O)OP(=O)([O-])[O-].[Ca+2].[Ca+2]

Specifications

Color White
Quantity 100 g
Solubility Information Soluble in water
Formula Weight 418.24
Percent Purity 95%
Physical Form Powder
Chemical Name or Material D-Fructose-1,6-diphosphate dicalcium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.