accessibility menu, dialog, popup

UserName

CaCaOH(S)OPOOO(S)OH(R)OHOOPOOO

CAS RN 6055-82-9

IUPAC Name: dicalcium (3R,4S,5S)-3,4,5-trihydroxy-2-oxo-6-(phosphonatooxy)hexyl phosphate
Synonyms: dicalcium (3R,4S,5S)-3,4,5-trihydroxy-2-oxo-6-(phosphonatooxy)hexyl phosphate
Molecular Weight (g/mol): 416.24
Molecular Formula: C6H10Ca2O12P2
InChi Key: MKSZTTCFYSGQHA-XAIJJRKESA-J
SMILES: [Ca++].[Ca++].O[C@@H](COP([O-])([O-])=O)[C@H](O)[C@@H](O)C(=O)COP([O-])([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)