missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 8-Specific Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | Cytokeratin 8-Specific |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17541356
|
Proteintech
27105-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17531356
|
Proteintech
27105-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Cytokeratin 8-specific antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human Cytokeratin 8-specific. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 8-specific antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Cytokeratin 8-Specific | |
Polyclonal | |
Rabbit | |
3856 | |
Primary |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human, Mouse, Rat | |
IgG |