Learn More
Cytokeratin 8-Specific Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27105-1-AP-20UL
Additional Details : Weight : 0.00020kg
 View more versions of this product
Product Code. 17541356
  • 1611.84 NOK / 20µL
Estimated Shipment: 08-05-2024
Description
The Cytokeratin 8-specific antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human Cytokeratin 8-specific. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 8-specific antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
Cytokeratin 8-Specific
Polyclonal
Rabbit
Primary
Human, Mouse, Rat
Western Blot, Immunohistochemistry, Immunofluorescence
Unconjugated
20 μL
3856