Learn More
Cytokeratin 8-Specific Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27105-1-AP-20UL
Additional Details : Weight : 0.00020kg
Description
The Cytokeratin 8-specific antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human Cytokeratin 8-specific. This antibody recognizes human, mouse, rat antigen. The Cytokeratin 8-specific antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Cytokeratin 8-Specific | |
Polyclonal | |
Rabbit | |
Primary | |
Human, Mouse, Rat |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
20 μL | |
3856 | |
IgG |