Learn More
R&D Systems™ Cynomolgus Monkey PVRIG (EHH62181) VersaClone cDNA
Specifications
Specifications
| Accession Number | EHH62181 |
| Gene ID (Entrez) | 79037 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date |
| Species | Cynomolgus Monkey |
| Product Line | R&D Systems |
| Quantity | 10 μg |