Learn More
R&D Systems™ Cynomolgus Monkey LYAG/GAA (XP_005585247.1) VersaClone cDNA
Specifications
Specifications
| Gene | LYAG/GAA |
| Accession Number | XP_005585247.1 |
| Gene ID (Entrez) | 2548 |
| Gene Alias | Acid alpha-Glucosidase, Acid maltase, Aglucosidase alfa, EC 3.2.1.20, GAA, glucosidase, alpha; acid, LYAG, lysosomal alpha-glucosidase, Lysosomal alphaGlucosidase |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |