Learn More
R&D Systems™ Cyno Monkey Osteoactivin (XP_005550047.1) VersaClone cDNA
missing translation for 'orderingAttributeHoverText'
Quantity:
10 μg
missing translation for 'unitSize'
10µg
Specifications
Specifications
| Gene | Osteoactivin/GPNMB |
| Accession Number | XP_005550047.1 |
| Gene ID (Entrez) | 10457 |
| Gene Alias | DC-HIL, glycoprotein (transmembrane) nmb, glycoprotein NMB, GPNMB, HGFIN, HGFINglycoprotein nmb-like protein, NMBosteoactivin, Transmembrane glycoprotein HGFIN, transmembrane glycoprotein NMB |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Quantity | 10 μg |