Learn More
R&D Systems™ Cyno Monkey IL-38/IL-1F10 (XP_005575360) VersaClone cDNA
Specifications
Specifications
| Accession Number | XP_005575360 |
| Gene ID (Entrez) | 84639 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Cynomolgus Monkey |
| Product Line | R&D Systems |
| Quantity | 10 μg |