Learn More
Cytiva CyDye™ DIGE Fluor Minimal Dye Labeling Kits
Product Code. 10066084 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
2 nmol of each: Cy2, Cy3, and Cy5
5 nmol of each: Cy2, Cy3, and Cy5
Unit Size:
1 set
Each
Product Code. Quantity unitSize
10066084 2 nmol of each: Cy2, Cy3, and Cy5 1 set
10279184 5 nmol of each: Cy2, Cy3, and Cy5 Each
2 options
Product Code. 10066084
Brand: Cytiva 28934530
Allow detection of up to three prelabeled protein samples and standards on the same 2-D electrophoresis gel. Amersham CyDye™ DIGE Fluor Minimal Labeling Kits contain size- and charge-matched fluorescent dyes specifically designed for detecting protein abundance differences in 2-D Fluorescence Difference Gel Electrophoresis using Ettan DIGE system.
  • Size- and charge-matched dyes enable co-migration of labeled samples within the gel
  • Bright and highly sensitive dyes allow the use of the minimal labeling technique
  • Minimal loss of signal during labeling, separation, and scanning
  • No change in signal over wide pH range used during first-dimension (IEF) separation
  • Discrete signal from each fluor with minimal cross-talk contributes to high accuracy
All minimal dyes allow the labeling of 50μg protein per labeling reaction. Minimal Dye Labeling Kits contain 2nmol or 5nmol of each of the minimal dyes.
For general 2-D applications where sufficient amounts of sample are available.
TRUSTED_SUSTAINABILITY
Specifications
Format Solid
For Use With (Application) The Scarce Sample Labeling Kit is designed to be used for applications in which only small amounts of sample are available, for example, in laser capture microdissection, detects up to 3 prelabeled protein samples, standard on the same 2-D electrophoresis gel
Quantity 2 nmol of each: Cy2, Cy3, and Cy5
Product Type Amersham CyDye DIGE Fluor Minimal Dye Labeling Kit
Kit Contents 2nM of CyDye DIGE Fluor minimal labeling kit (Cy2, Cy3 and Cy5)
Content And Storage -15°C to -30°C
Color Blue, Green, Red
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Cy2, Cy3, Cy5