Learn More
Fisherbrand™ Customizable Foam Stand for Vortex Mixer
Product Code. 11792573 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Diameter (Metric):
94 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Unit Size
11792573 94 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11792573
  • ONLINE EXCLUSIVE PRICE
    320.00 NOK
    314.24 NOK / Pack of 1
    Save 5.76 NOK
    1% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
Securely hold test tubes and vials in the configuration that works best for your needs. Customizable Foam Stands are compatible with Vortex Mixers.
  • Soft foam to prevent breakage
  • Black in color
Specifications
Diameter (Metric) 94 mm
For Use With (Equipment) CLASSIC, ZX3, WIZARD, ZX4, TX4