missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPSF4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15023-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
LUZP1 encodes a protein that contains a leucine zipper motif. The exact function of the encoded protein is not known. In mice this gene affects neural tube closure.Specifications
CPSF4 | |
Polyclonal | |
Unconjugated | |
CPSF4 | |
CPSF 30 kDa subunit, CPSF30, CPSF4, NAR, Neb 1, NEB1, No arches homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10898, 54188 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O95639, Q8BQZ5 | |
Cpsf4 | |
CPSF4 Fusion Protein Ag7103 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |