missing translation for 'onlineSavingsMsg'
Learn More

CPSF4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. p-7207096 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16883994 20 μL 20µL
16873994 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16883994 Supplier Proteintech Supplier No. 150231AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LUZP1 encodes a protein that contains a leucine zipper motif. The exact function of the encoded protein is not known. In mice this gene affects neural tube closure.

Specifications

Antigen CPSF4
Applications Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CPSF4
Gene Accession No. O95639, Q8BQZ5
Gene Alias CPSF 30 kDa subunit, CPSF30, CPSF4, NAR, Neb 1, NEB1, No arches homolog
Gene Symbols Cpsf4
Host Species Rabbit
Immunogen CPSF4 Fusion Protein Ag7103
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10898, 54188
Target Species Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.