Learn More
Corning™ 96-Well, Non-Treated, Flat-Bottom, Half-Area Microplate
GREENER_CHOICE
Product Code. 10634102 Shop All Corning Products
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 100
Product Code. Color unitSize
10634102 Black Pack of 100
1 options
Product Code. 10634102
Brand: Corning™ 3694
Only null left
Add to basket
Add to basket
Color:Black; Sterility:Non-sterile; Lid:Without Lid
  • Designed to reduce well-to-well cross-talk
  • White microplates enhance luminescent signals and have low background luminescence.
  • Black microplates have low background fluorescence and minimize light scattering.
  • TRUSTED_SUSTAINABILITY
    Specifications
    Barcode Alphanumeric
    Format Half-Area
    Lid Without Lid
    Array 8 x 12
    Color Black
    For Use With (Equipment) Plate readers
    Material Polystyrene
    No. of Wells 96
    Volume (Metric) Working 25 to 125 μL
    Sterility Non-sterile
    Surface Treatment Non-treated
    Well Shape Flat Bottom (F-well)
    For Use With (Application) Suitable for immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
    No. per Case 100/Cs.
    No. per Pack 25/Pk.
    Quantity 100/Cs.