Specifications
| Shape | Round |
| For Use With (Application) | Flask Support |
| Quantity | 1 Each |
| Depth (Metric) | 38 mm |
| Diameter (Metric) Outer | 150 mm |
| Product Type | Cork Ring |
| Diameter (Metric) Inner | 120 mm |